Learn More
BD Cytometric Bead Array (CBA) Mouse Enhanced Sensitivity Master Buffer Kit
For use in Flow Cytometry
Brand:  BD 562246
 View more versions of this product
Product Code. 15861749
  • £1230.00 / Each
Estimated Shipment: 29-11-2024
to see stock.
Explore more special offers
Add to basket
Includes: Assay Diluent (150mL), Capture Bead Diluent (30mL), Detection Reagent Diluent A (15mL), Detection Reagent Diluent B (35mL), Wash Buffer (650mL)
Description
Description
Host Species:
Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Mouse
100 Tests
Flow Cytometry
Assay diluent 150mL, capture bead Diluent 30mL, detection reagent diluent A 15mL, detection reagent diluent B 35mL, Wash Buffer 650mL
2°C to 8°C
Mouse enhanced sensitivity master Buffer kit