missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Cytometric Bead Array (CBA) Mouse Enhanced Sensitivity Master Buffer Kit
For use in Flow Cytometry
£1230.00 - £1765.00
Specifications
Cross Reactivity | Mouse |
---|---|
Content And Storage | 2°C to 8°C |
For Use With (Application) | Flow Cytometry |
Product Type | Mouse enhanced sensitivity master Buffer kit |
Includes: Assay Diluent (150mL), Capture Bead Diluent (30mL), Detection Reagent Diluent A (15mL), Detection Reagent Diluent B (35mL), Wash Buffer (650mL)
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15881749
|
BD
562248 |
500 Tests |
£1765.00
Each |
Estimated Shipment: 29-11-2024
Log in to see stock. |
|||||
15861749
|
BD
562246 |
100 Tests |
£1230.00
Each |
Estimated Shipment: 29-11-2024
Log in to see stock. |
|||||
Description
Host Species:
Flow Cytometry
Specifications
Mouse | |
2°C to 8°C | |
Flow Cytometry | |
Mouse enhanced sensitivity master Buffer kit |