missing translation for 'onlineSavingsMsg'
Learn More

BD Cytometric Bead Array Human IP-10 Flex Set Standard

Lyophilized human IP-10

Brand:  BD 558466


Product Code. 15889408

  • £163.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Lyophilized in an aqueous buffered solution containing BSA and ProClin™ 150
  • Protein concentration of 25,000pg/ml (when reconstituted in 0.4ml Assay Diluent)
  • Regulatory status: RUO
  • Recombinant

Flow cytometry, controls

Order Info

100 tests.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Aqueous Buffered Solution containing BSA and ProClin™ 150
100 Tests
Flow cytometry
25000pg/mL
0.2mL
Lyophilized
Technical data sheet
Cytometric Ab Control Standards
RUO
4°C
Human
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Human IP-10 Standard

Signal Word
  • Warning

Hazard Category
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : CMIT/MIT

SDS
Documents

Documents

Certificates
Special Offers

Special Offers