missing translation for 'onlineSavingsMsg'
Learn More

BD Cytometric Bead Array Mixture

Control sample to demonstrate proper setup of flow cytometer and FCAPArray™ software

Brand:  BD 558522

Additional Details : Weight : 0.01000kg

Product Code. 15829458

  • £249.00 / Each
Estimated Shipment: 02-08-2024
to see stock.

Alert:

Dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing


Description

Description

The bead mixture contains bead positions A4, A5, A6, A7, A8, A9, B4, B5, B6, B7, B8, B9, C4, C5, C6, C7, C8, C9, D4, D5, D6, D7, D8, D9, E4, E5, E6, E7, E8 and E9. It is only a guarantee that the instrument and FCAP Array Software (Cat. No. 641488) can analyze the 30 plex correctly

  • 30 Plex Bead Mixture is a cocktail
  • Contains 30 bead populations from BD CBA Flex Set
  • Has distinct fluorescence intensities resolvable in NIR and Red channels of BD FACSArray bioanalyzer
  • Storage Buffer: Aqueous buffered solution containing fetal bovine serum and ≤0.09% sodium azide

Array

Specifications

Specifications

Aqueous buffered solution containing fetal bovine serum and ≤0.09% sodium azide
25 μL
30 bead
Flow cytometer
4°C
10
Flow Cytometry Calibration and Alignment
RUO
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Research use only