Learn More
BD D- Luciferin Sodium Salt
Specific for firefly luciferase in the illustrated reactions
Brand:  BD 556876
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 15848788
  • £1065.00 / Each
Estimated Shipment: 29-08-2024
to see stock.
Add to basket
Includes: Sodium salt
Description
Description
Purified by HPLC techniques to remove dehydroluciferin and L-Luciferin, both of which are competitive inhibitors of firefly luciferase. Form dehydroluciferin in presence of ultraviolet light and/or moisture
  • Synthetically made substrate
  • lyophilized, in monosodium salt form
  • Non-buffered solution
  • Reconstitute lyophilized D-Luciferin with deionized water
Specifications
Specifications
Lyophilized
D-Luciferin
-20°C
Sodium salt
100 mg
Luciferase assay
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Research use only