missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD D- Luciferin Sodium Salt
Specific for firefly luciferase in the illustrated reactions
£699.00 - £1065.00
Specifications
Formulation | Lyophilized |
---|---|
Kit Contents | Sodium salt |
Type | D-Luciferin |
Content And Storage | -20°C |
For Use With (Application) | Luciferase assay |
Includes: Sodium salt
Description
Purified by HPLC techniques to remove dehydroluciferin and L-Luciferin, both of which are competitive inhibitors of firefly luciferase. Form dehydroluciferin in presence of ultraviolet light and/or moisture
- Synthetically made substrate
- lyophilized, in monosodium salt form
- Non-buffered solution
- Reconstitute lyophilized D-Luciferin with deionized water
Specifications
Lyophilized | |
D-Luciferin | |
Luciferase assay |
Sodium salt | |
-20°C |
Research use only