Learn More
BD Difco™ Dehydrated Culture Media: Granulated Agar
Used as a solidifying agent for culture media. BD Difco™ Dehydrated Culture Media: Granulated Agar is carefully monitored for cultural response, solubility and gelation temperature.
Brand:  BD 214510
 View more versions of this product
Product Code. 11718233
  • £692.00 / 2kg
Estimated Shipment: 16-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Applications:
  • Solidifying agent for culture media
  • Granulated
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ISO/DIS, 11014
Agar
General bacteriological purposes
2000 g
1.5%
Dehydrated Granule
Container