missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Granulated Agar
Used as a solidifying agent for culture media. BD Difco™ Dehydrated Culture Media: Granulated Agar is carefully monitored for cultural response, solubility and gelation temperature.
£566.00 - £692.00
Specifications
Certifications/Compliance | ISO/DIS, 11014 |
---|---|
Concentration | 1.5% |
Additive | Agar |
Form | Dehydrated Granule |
For Use With (Application) | General bacteriological purposes |
Description
Applications:
- Solidifying agent for culture media
- Granulated
Specifications
ISO/DIS, 11014 | |
Agar | |
General bacteriological purposes |
1.5% | |
Dehydrated Granule | |
Container |
Safety and Handling
missing translation for 'shelfLife' : 1825 Days
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BD Difco™ Dehydrated Culture Media: Granulated Agar