Learn More
BD Difco™ Dehydrated Culture Media: Marine Broth 2216
Cultivates heterotrophic marine bacteria. BD Difco™ Dehydrated Culture Media: Marine Broth 2216 has a shelf life of 1825 days.
Brand: BD 279110
Description
- Dehydrated
- Culture Media
- Marine Broth 2216

Specifications
CE | |
Dehydrated | |
Microbiology Media | |
Cultivation of heterotropic marine bacteria | |
500 g |
Refrigerate | |
Marine Broth 2216 | |
Peptone 5g/L, Yeast extract 1g/L, Ferric Citrate 0.1g/L, Sodium Chloride 19.45g/L, Magnesium Chloride 5.9g/L, Magnesium Sulfate 3.24g/L, Calcium Chloride 1.8g/L, Potassium Chloride 0.55g/L, Sodium Bicarbonate 0.16g/L, Potassium Bromide 0.08g/L, Strontium | |
Bottle |
Safety and Handling
- marine broth 2216
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : calcium chloride
Your input is important to us. Please complete this form to provide feedback related to the content on this product.