Learn More
BD GasPak™ EZ Gas Generating Systems and Supplies: Incubation Racks
Catalyst-free convenience for use in producing anaerobic, microaerophilic, or CO2-enriched environments
Brand: BD 260674
This item is not returnable.
View return policy
Description
Catalyst-free convenience for use in producing anaerobic, microaerophilic, or CO2-enriched environments
Holds 30 to 33 petri dishes.
Specifications
EZ Incubation Rack | |
Petri Dishes (30 to 33) | |
Each |
Primary isolation and cultivation of anaerobic, microaerophilic or capnophilic bacteria by use of gas generating sachets | |
GasPak | |
Large |
Product Content Correction
Product Title
BD GasPak™ EZ Gas Generating Systems and Supplies: Incubation Racks > Large Incubation Container Rack
Spot an opportunity for improvement?Share a Content Correction