Learn More
BD GasPak™ Supplies
Used with the GasPak™ EZ Gas Generating Systems. BD™ GasPak™ Supplies are used with these complete, integrated systems for rapid, waterless and catalyst-free gas generation. Can be used to support an anaerobic, microaerophilic or CO2 enriched environment.
Brand:  BD 271051
Code : 03
Additional Details : Weight : 0.03330kg
 View more versions of this product
Product Code. 11703473
  • £743.00 / Pack of 100
Estimated Shipment: 23-08-2024
to see stock.
Add to basket
Description
Description
GasPak™ EZ Gas Generating Systems are complete, integrated systems that rapidly produce atmospheres suitable to support the primary isolation and cultivation of anaerobic, microaerophilic or capnophilic bacteria by use of gas generating sachets inside multi-use incubation containers.
Accessories include:
  • CO2 Indicator Strips
  • Dry Anaerobic Indicator Strips
  • Tube Holder
  • Catalyst Replacement Charges
  • Catalyst Reaction Chamber
Specifications
Specifications
Dry Anaerobic Indicator Strip
Strip changes from blue to colorless in the absence of oxygen.
Produce atmospheres suitable to support the primary isolation and cultivation of anaerobic, microaerophilic or capnophilic bacteria
100 Pk.
15°C to 30°C
Strip
BD GasPak™