Learn More
BD GasPak™ EZ Gas Generating Systems and Supplies: Pouch Systems
Used in producing anaerobic, microaerophilic, or CO2-enriched environments. BD Difco™ GasPak™ EZ Gas Generating Systems and Supplies provide catalyst-free convenience.
Brand: BD 260685
Description
- One-step resealable pouches
- The pouches are specially designed to maximize the preservation of the desired environment throughout incubation
- This heat-free technology minimizes condensate, allowing excellent bacterial colony visualization though the pouch
Specifications
EZ CampyPouch System | |
Resealable Pouches | |
BD GasPak EZ Small Incubation Container | |
GasPak |
Ziplock | |
Primary isolation and cultivation of anaerobic, microaerophilic or capnophilic bacteria by use of gas generating sachets | |
Resealable pouches | |
20 Sachets |