Learn More
BD Fixable Viability Stain 575V
For discrimination of viable from non-viable mammalian cells in multicolor flow cytometric applications. BD Horizon™ Fixable Viability Stain 575V reacts with and covalently binds to cell-surface and intracellular amines. Permeable plasma cell membranes allow for the intracellular diffusion of the dye and covalent binding to higher overall concentrations of amines than in nonpermeable live cells.
Brand: BD 565694
209.04 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Fixable Viability Stain 575V is excited by the Violet laser (with an excitation maximum of 396nm), and has a fluorescence emission maximum of 572nm

Specifications
-80°C | |
|
|
Fixable Viability Stain 575V |
200 μg | |
Permeable |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- BD Horizon Fixable Viability Stain 575V
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Fixable Viability Stain 575V
Your input is important to us. Please complete this form to provide feedback related to the content on this product.