Learn More
BD BV480 Mouse Anti-Human CD4
CD4 distribution on lymphocytes is similar for both human and monkey cells. BD Horizon™ BV480 Mouse Anti-Human CD4, with the majority of CD4-positive lymphocytes being CD8-negative and lacking reactivity with antibodies to B- or NK-cell markers.
Brand: BD 566159
Additional Details : Weight : 0.00900kg
Description
- With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set
- BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
Specifications
BV480 Mouse Anti-Human CD4 | |
Flow Cytometry | |
BV480 | |
Store undiluted at 4°C |
25 Tests | |
QC Testing: Rhesus, Cynomolgus, Baboon; Tested in Development: Human | |
5 μL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide |