missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BV480 Mouse Anti-Human CD4
CD4 distribution on lymphocytes is similar for both human and monkey cells. BD Horizon™ BV480 Mouse Anti-Human CD4, with the majority of CD4-positive lymphocytes being CD8-negative and lacking reactivity with antibodies to B- or NK-cell markers.
£294.00 - £585.00
Specifications
Product Type | BV480 Mouse Anti-Human CD4 |
---|---|
For Use With (Application) | Flow Cytometry |
Cross Reactivity | QC Testing: Rhesus, Cynomolgus, Baboon; Tested in Development: Human |
Form | BV480 |
Sample Volume | 5 μL |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15834789
|
BD
566148 |
100 Tests |
£585.00
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
15844799
|
BD
566159 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set
- BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
Specifications
BV480 Mouse Anti-Human CD4 | |
QC Testing: Rhesus, Cynomolgus, Baboon; Tested in Development: Human | |
5 μL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide |
Flow Cytometry | |
BV480 | |
Store undiluted at 4°C |