missing translation for 'onlineSavingsMsg'
Learn More

BD BV480 Mouse Anti-Ki-67

Expressed in the nucleus of cycling cells (G1, S, G2, M cell cycle phases). BD Horizon™ BV480 Mouse Anti-Ki-67 is a large protein having 2 alternatively spliced isoforms, an N-terminal forkhead-associated domain, a C-terminal domain that binds to heterochromatin proteins, and multiple phosphorylation sites, the functions of which are still unclear.

Brand:  BD 566109

 View more versions of this product

Product Code. 15874749

  • £605.00 / Each
Estimated Shipment: 10-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set
  • BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
TRUSTED_SUSTAINABILITY
Specifications

Specifications

BV480 Mouse Anti-Ki-67
Intracellular Staining (Flow Cytometry)
BV480
Store undiluted at 4°C
100 Tests
QC Testing: Human; Tested in Development: Mouse; Reported Reactivity: Rat, Rhesus
5 μL
Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers