missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BV480 Mouse Anti-Ki-67
Expressed in the nucleus of cycling cells (G1, S, G2, M cell cycle phases). BD Horizon™ BV480 Mouse Anti-Ki-67 is a large protein having 2 alternatively spliced isoforms, an N-terminal forkhead-associated domain, a C-terminal domain that binds to heterochromatin proteins, and multiple phosphorylation sites, the functions of which are still unclear.
£325.00 - £605.00
Specifications
Product Type | BV480 Mouse Anti-Ki-67 |
---|---|
For Use With (Application) | Intracellular Staining (Flow Cytometry) |
Cross Reactivity | QC Testing: Human; Tested in Development: Mouse; Reported Reactivity: Rat, Rhesus |
Form | BV480 |
Sample Volume | 5 μL |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15874809
|
BD
566172 |
25 Tests |
£325.00
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
15874749
|
BD
566109 |
100 Tests |
£605.00
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
Description
- With an Ex Max of 436nm and Em Max at 478nm, BD Horizon™ BV480 can be excited by the violet laser and detected in the BD Horizon™ BV510 (525/40nm) filter set
- BV480 has less spillover into the BV605 detector and, in general, is brighter than BV510
Specifications
BV480 Mouse Anti-Ki-67 | |
QC Testing: Human; Tested in Development: Mouse; Reported Reactivity: Rat, Rhesus | |
5 μL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide |
Intracellular Staining (Flow Cytometry) | |
BV480 | |
Store undiluted at 4°C |