missing translation for 'onlineSavingsMsg'
Learn More

BD ICC fixation buffer

For cytokine immunocytochemical staining

Brand:  BD 550010

198.57 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 15826168

  • £201.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

Contains formalin, which is harmful and a suspected carcinogen

This item is not returnable. View return policy

Description

Description

Useful for fixation of unstained cells adhered to microscopic slides

  • Neutral pH-buffered saline
  • Used to preserve human and rodent lymphoid cells

Immunocytochemistry (cytospins)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

4°C
Fixation Buffer
100mL
Fixing unstained cells on slides for Cytokine immunocytochemical staining
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BD ICC fixation buffer > ICC Fixation Buffer; 100mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Research Use Only