missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD PBX Calcium Assay Kit
For homogeneous measurement of intracellular calcium changes
£766.00 - £5435.00
Specifications
For Use With (Application) | For homogeneous measurement of intracellular calcium changes |
---|---|
Type | Calcium Assay Kit |
Includes: Calcium Indicator, 10X PBX Signal Enhancer
Product Code | Brand | Includes | Quantity | Content And Storage | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Quantity | Content And Storage | Price | Quantity & Availability | |||
15896279
|
BD
640175 |
1 vial calcium indicator,10mL PBX signal enhancer (10X, lyophilized), 90mL calcium assay buffer 1X | 10 Preps | -20°C |
£766.00
1 set |
Estimated Shipment: 18-10-2024
Log in to see stock. |
|||
15816289
|
BD
640177 |
10 vials calcium indicator,100mL PBX signal enhancer (10X, lyophilized) | 100 Preps | Calcium indicator at -20°C and PBX Signal Enhancer 10X at room temperature |
£5435.00
1 set |
Estimated Shipment: 22-10-2024
Log in to see stock. |
|||
Description
- Involves one dye addition step
- Does not require washing
- Allows easy implementation in highthroughput environment
- Probenecid, a standard inhibitor of nonspecific anion transport, is not required for dye-loading
- Can improve retention of the dye in cells
Specifications
For homogeneous measurement of intracellular calcium changes | |
Calcium Assay Kit |
Research use only