Learn More
BD DRAQ5™
Used to stain nucleic acids in live or fixed cells. BD Pharmingen™ DRAQ5™ is also used for cell cycle analysis by DNA content, nuclear visualization, or discrimination of nucleated cells from debris or enucleated cells.
Brand:  BD 564903
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15803959
  • £677.00 / Each
Estimated Shipment: 18-10-2024
to see stock.
Description
Description
  • Far-red fluorescent DNA dye
  • Cell permeable
  • Used to stain nucleic acids in live or fixed cells
  • Has an excitation wavelength maxima of 598/646nm
Specifications
Specifications
Far-red
Store undiluted at 4°C
DNA dye
50 μL
5 mM
Bioimaging, Immunofluorescence, Intracellular Staining (Flow Cytometry)
598/646 nm
Tested in Development: Human, Mouse; Reported Reactivity: Rat