missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Recombinant Human β2 Microglobulin
Recombinant protein expressed in insect cells and purified by a combination of ion-exchange and gel-filtration methods
Brand: BD 551089
Additional Details : Weight : 0.01000kg
Description
- Storage buffer: Aqueous buffered solution containing ≤0.09% sodium azide
- Maximal activity range: 50-200ng/mL
Specifications
4°C | |
Beta-2 Microglobulin | |
0.1 mg |
Research use only.