Learn More
Benchmark Scientific™ BeadBath Duo™
The BeadBath Duo allows for precise temperature control when using either water or thermal beads.
£886.00
Specifications
For Use With (Equipment) Tubes, Bottles, Flasks
Temperature Range 5°C to 80°C
Type Bath
Voltage 230 V
Frequency 50/60 Hz
View More Specs
Products 1
Product Code Brand Capacity (Metric) Dimensions (D x W x H) Dimensions (D x W x H) Interior Price Quantity & Availability  
Product Code Brand Capacity (Metric) Dimensions (D x W x H) Dimensions (D x W x H) Interior Price Quantity & Availability  
17997070
View Documents
Benchmark Scientific™
B2402-E
2 L 20.3 x 16.5 x 15.2 cm 15.2 x 13.9 x 10.1 cm
£886.00
Each
Description
Description
  • Convert from a water bath to a bead bath at the touch of a button
  • Two separate modes for beads or water ensure optimal performance
  • Quik-Cal™, simple in-lab calibration function
  • Accurate and uniform temperature control
  • No constant refilling
  • Accessory-free securing of containers
  • Ability to run constantly without waiting for heating
Specifications
Specifications
Tubes, Bottles, Flasks
Bath
50/60 Hz
5°C to 80°C
230 V