Learn More
Bel-Art™ SP Scienceware™ Acrylic Desiccator Cabinets
Used in refrigerators or at room temperature
Brand: Bel-Art™ F42066-0001
This item is not returnable.
View return policy
Description
- Acrylic cabinet with removable, tray-like shelves with perforated bottoms and slide in/out on pre-set rails
- Door opens from right to left and secured with stainless steel latch
- Available with three shelves
- Comes in clear color and bronze color for light-sensitive materials
Specifications
Bronze | |
Gasketed | |
32 Cans | |
3 removable tray-like shelves | |
12 x 12 in. (30.4 x 30.4 cm) | |
30.4 cm | |
30.4 cm | |
30.4 cm |
Acrylic | |
Used in refrigerators/at room temperature for light-sensitive materials | |
Tray-like 3 shelves with perforated bottoms and slide in/out on pre-set rails, stainless steel twist locking latch for door | |
0.82 cu. ft. | |
12 in. | |
12 in. | |
12 in. |