missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Bacterial Cell Spreaders

Spreads bacterial cells onto the surface of an agar Bel Art™ SP Scienceware™ Bacterial Cell Spreaders are available in two sizes.

Brand:  Bel-Art™ F37736-0006

 View more versions of this product

Product Code. 11326954

  • £34.95 / Each
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Spreads bacterial cells onto the surface of an agar Bel Art™ SP Scienceware™ Bacterial Cell Spreaders are available in two sizes.

  • Autoclavable and may be flame sterilized
  • Features triangular end with smooth rounded surfaces throughout
  • Smaller 16mm width bar suitable for use with 6cm plates
  • Larger 25mm width bar suitable for use with 9cm plates
  • Use with Inoculating Turntable (Mfr. No.F40913-0000)
  • Both sizes 30cm long
Specifications

Specifications

Autoclavable
12 in.
Stainless Steel
Sterile
Inoculating turntable, 6cm inoculating plates
30.2 cm
Bacterial Cell Spreader
Easily perform inoculation of petri dishes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers