missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Clear Graduated Cylinders

No concave meniscus, makes the raised numeral, single scale, graduations simple to interpret.

Brand:  Bel-Art™ F28690-0000

 View more versions of this product

Product Code. 30193423

  • £8.35 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Made from clear TPX
  • Shatter-proof
  • Wide octagonal base prevents tipping and rolling
  • Chemical and heat resistant to 177°C (350°F)
  • Steam autoclavable at 121°C (250°F)

Specifications

Specifications

10 mL
Chemical and heat resistant
0.1mL
Clear TPX Plastic
0.1 mL
Graduated Cylinder, 10 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.