Learn More
Bel-Art™ SP Scienceware™ Clear Graduated Cylinders
No concave meniscus, makes the raised numeral, single scale, graduations simple to interpret.
£8.35 - £63.40
Product Code | Brand | Graduations | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Graduations | Type | Price | Quantity & Availability | ||||
30193476
|
Bel-Art™
F28691-0000 |
0.5 mL | Graduated Cylinder, 25 mL |
£10.60
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
30193423
|
Bel-Art™
F28690-0000 |
0.1 mL | Graduated Cylinder, 10 mL |
£8.35
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17824728
|
Bel-Art™
F28692-0000 |
1 mL | Graduated Cylinder, 50 mL |
£11.64
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17834728
|
Bel-Art™
F28694-0000 |
2 mL | Graduated Cylinder, 250 mL |
£13.47
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
30193477
|
Bel-Art™
F28693-0000 |
1 mL | Graduated Cylinder, 100 mL |
£13.30
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17844728
|
Bel-Art™
F28695-0000 |
5 mL | Graduated Cylinder, 500 mL |
£18.02
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
30193431
|
Bel-Art™
F28696-0000 |
10 mL | Graduated Cylinder, 1000 mL |
£28.15
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
30193436
|
Bel-Art™
F28697-0000 |
20 mL | Graduated Cylinder, 2000 mL |
£63.40
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Made from clear TPX
- Shatter-proof
- Wide octagonal base prevents tipping and rolling
- Chemical and heat resistant to 177°C (350°F)
- Steam autoclavable at 121°C (250°F)
Specifications
Chemical and heat resistant |