missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ Clear View Fume Hood
Safely vent noxious or potentially hazardous fumes and dust with the Bel-Art™.
Brand: Bel-Art™ F50020-1010
2075.75 GBP valid until 2025-04-30
Use promo code "24235" to get your promotional price.
Includes: Laboratory, classroom or industrial
Description
- Versatile 3-position sash
- Clear polycarbonate provides optimum visibility and durability
- 500201010, Top sitting: 6 in. O.D. vent
- O.D.: 24D x 25.8W x 26.12 in. H (61 x 65.7 x 66.4 cm)
- I.D.: 20D x 24W x 24 in. H (50.8 x 61 x 61 cm)
- Internal Space: 11.67 cu. ft. (0.18 m)
- 500202010, Two Top sitting: 6 in. O.D. vent
- O.D.: 24D x 43.8W x 26.1 in. H (61 x 111.4 x 66.4cm)
- I.D.: 20D x 42W x 24 in. H (50.8 x 106.7 x 61 cm)
- Internal Space: 11.67 cu. ft. (0.18 m)
Specifications
Fume Hood | |
Unfiltered | |
Polycarbonate | |
61 cm | |
26 in. | |
25.87 in. |
Uncoated | |
None | |
24 in. | |
65.7 x 61.0 x 66.3 cm | |
Laboratory, Classroom | |
65.7 cm |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Multiple Manufacturers
Equipment Monthly Promotion
Discover exclusive monthly offers on scientific equipment to enhance your laboratory's capabilities.Promo Code: 24235Expires: 2025-04-30
Days
27Hours
15Minutes
14Seconds
10New Offer Save Up to 50%
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Bel-Art™ Clear View Fume Hood > 1 top-sitting 6 in. O.D. vent opening; I.D.: 20D x 24W x 24 in. H; O.D.: 24D x 25.8W x 26.12 in. H
Spot an opportunity for improvement?Share a Content Correction