missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Labeled Wash Bottles
Polyethylene bottles with polypropylene closures to ensure leakproof dispensing.
£32.31 - £49.00
Specifications
Material | LDPE, Polyethylene |
---|---|
Autoclavable | Not Autoclavable |
Closure Material | Polypropylene |
Closure Size | 53 mm |
Alert:
All bottles are intended for dispensing use only, not for long-term storage. Use LYOB only with solvents compatible with LDPE.
Includes: Safety labeled wash bottles include Department of Transportation (DOT) and National Fire Protection Association (NFPA) codes and symbols, Chemmical Abstract Service (CAS), protective equipment symbols and target organ information.
Product Code | Brand | Capacity | For Use With (Application) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | For Use With (Application) | Price | Quantity & Availability | ||||
11592463
|
Fisherbrand™
00622-1500 |
16 oz., 500 mL | Acetone | N/A | |||||
30193464
|
Fisherbrand™
F11646-0624 |
16 oz., 500 mL | Isopropanol |
£49.00
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11572463
|
Fisherbrand™
00621-1500 |
16 oz., 500 mL | Water | N/A | |||||
Description
- Easy-to-read printed bottles
- Wide mouth 53mm (2 in.) bottle for easy refilling
- Translucent, LDPE solvent-resistant
- Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm
- Toluene is red LDPE bottle
- Polypropylene closure with leakproof design
- Bottles can be purchased separately or in variety packs that include water, acetone, methanol, isopropanol, toluene and ethanol
Compliance
OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
Specifications
LDPE, Polyethylene | |
Not Autoclavable | |
Polypropylene | |
53 mm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title