missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Mini Magnifier

Provides hands-free 1.75X magnification of slides, plates or gels placed on the Mini Light Box II

Brand:  Bel-Art™ F37865-0000

Product Code. 10216204

  • £276.04 / Each
Estimated Shipment: 27-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Tilts for optimal viewing
  • Measures 10.5 x 14.3 x 17.2cm H
  • Also useful for viewing of x-ray films, stained gels, slides or text
Specifications

Specifications

Magnified observation of petri dishes, slides, gels or text
6.7 in.
6.25 in.
13 oz.
5.9 in.
5.5 in.
1.75x
Magnifier, Mini
Scienceware™ Mini Light Box II
17.2 cm
15.8 cm
368.5 g
14.9 cm
140 mm
Glass Lens
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bel-Art™ Mini Magnifier > Mini Magnifier

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.