missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Poxygrid™ Contact Plate/Petri Dish Racks

For safe transport, incubation and storage of contact plates and convenient organization

Brand:  Bel-Art™ F18979-0003

 View more versions of this product

Product Code. 13415399

  • £39.35 / Each
Estimated Shipment: 19-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Durable blue epoxy-coated steel wire rack holds contact plates with lids of 72mm maximum diameter
  • Chemical and corrosion resistant
  • Suited for RODAC™ and BD™ models
  • Compact size maximizes incubator space
  • Racks are steam autoclavable at 121°C (250°F)

Specifications

Specifications

Autoclavable
Hold 10 contact plates with lids (72mm max dia.)
10
3.25 x 3.25 x 5.5 in. (8.2 x 8.2 x 14cm)
9.62 in.
3.25 in.
5.5 in.
Blue
Steel Wire
Contact plates with lids
24.6 cm
8.2 cm
14 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.