missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Secador™ Auto Desiccator Cabinets: Horizontal Models

A UV light blocking, dry environment with automatic electronic dehumidification

Brand:  Bel-Art™ F42074-0226

 View more versions of this product

Product Code. 10096721

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

The attractive design and sturdy construction of the Secador line of desiccator cabinets assures easy and reliable storage of moisture and light sensitive products.

  • Patented electronic dehumidification system reduces humidity and circulates dry air automatically, freeing users from monitoring and changing desiccants
  • Durastar™ co-polyester construction blocks 99% of UV light and resists staining, crazing and chemical attack
  • Amber color models reduce visible light penetration by over 50% for storing light sensitive materials like reagents, analytical standards and investigational compounds
  • Large doors with easy-to-view dial hygrometer maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals
  • Perforated shelves facilitate circulation of dry air; Space for desiccant under bottom shelf
  • 4.0 horizontal has a wider footprint, lower height and pull down “oven style” door
  • U.S. Patents: 6,772,534; 6,834,920; 7,114,266; 7,318,630 and D474,626

  • 4.0 units can be converted from horizontal to vertical configuration or vice versa by turning the unit and ordering the appropriate number of shelves (see Ordering Info)
  • Dimensions
    • Secador 2.0: 1.16 cu. ft. internal volume, 16.3L × 13.4W × 12.4 in. H
    • Secador 3.0: 1.55 cu. ft. internal volume, 16.3L × 13.4W × 16.4 in. H
    • Secador 4.0: 1.87 cu. ft. internal volume, 16.3L × 13.4W × 20.4 in. H

    Order Info

    Replacement shelves are available: Replacement Shelf for Horizontal 4.0 Cabinets, Mfr. No. 420740001

    Specifications

    Specifications

    Not Autoclavable
    Horizontal
    Bottom-hinged, pull down oven style
    Carrying handles, 2 latches and loops for a padlock or tamper-evident seals, door-mounted hygrometer
    2
    Vented Adjustable
    5°C to 40°C
    20.4 in.
    51.8 cm
    13.4 in.
    34.1 cm
    41.4 cm
    Clear with Blue Ends
    Copolyester
    Easy and reliable storage of moisture and light sensitive products, can be used with Scienceware™ desiccant cartridges, canisters or similar desiccant
    4.0
    Chemicals, stains and fine cracks
    41°F to 104°F
    1.9 cu. ft.
    8.9 in.
    22.5 cm
    16.4 in.
    16.3 in.
    230 V
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.