missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Secador™ Auto Desiccator Cabinets: Horizontal Models
A UV light blocking, dry environment with automatic electronic dehumidification
£1095.00
Specifications
Autoclavable | Not Autoclavable |
---|---|
Cabinet Style | Horizontal |
Material | Copolyester |
Door Style | Bottom-hinged, pull down oven style |
For Use With (Application) | Easy and reliable storage of moisture and light sensitive products, can be used with Scienceware™ desiccant cartridges, canisters or similar desiccant |
Product Code | Brand | Color | No. of Shelves | Shelf Type | Voltage | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | No. of Shelves | Shelf Type | Voltage | Price | Quantity & Availability | ||
11842732
|
Bel-Art™
F42074-0220 |
Clear | 2 | Vented Adjustable | 230 V |
£1095.00
Each |
Estimated Shipment: 29-11-2024
Log in to see stock. |
||
11892732
|
Bel-Art™
F42074-0227 |
Blue with Clear Door | 2 | Vented Adjustable | 230 V |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
10096721
|
Bel-Art™
F42074-0226 |
Clear with Blue Ends | 2 | Vented Adjustable | 230 V |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
The attractive design and sturdy construction of the Secador line of desiccator cabinets assures easy and reliable storage of moisture and light sensitive products.
- Patented electronic dehumidification system reduces humidity and circulates dry air automatically, freeing users from monitoring and changing desiccants
- Durastar™ co-polyester construction blocks 99% of UV light and resists staining, crazing and chemical attack
- Amber color models reduce visible light penetration by over 50% for storing light sensitive materials like reagents, analytical standards and investigational compounds
- Large doors with easy-to-view dial hygrometer maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals
- Perforated shelves facilitate circulation of dry air; Space for desiccant under bottom shelf
- 4.0 horizontal has a wider footprint, lower height and pull down “oven style” door
- U.S. Patents: 6,772,534; 6,834,920; 7,114,266; 7,318,630 and D474,626
- 4.0 units can be converted from horizontal to vertical configuration or vice versa by turning the unit and ordering the appropriate number of shelves (see Ordering Info)
- Secador 2.0: 1.16 cu. ft. internal volume, 16.3L × 13.4W × 12.4 in. H
- Secador 3.0: 1.55 cu. ft. internal volume, 16.3L × 13.4W × 16.4 in. H
- Secador 4.0: 1.87 cu. ft. internal volume, 16.3L × 13.4W × 20.4 in. H
Order Info
Replacement shelves are available: Replacement Shelf for Horizontal 4.0 Cabinets, Mfr. No. 420740001
Specifications
Not Autoclavable | |
Copolyester | |
Easy and reliable storage of moisture and light sensitive products, can be used with Scienceware™ desiccant cartridges, canisters or similar desiccant | |
4.0 | |
Chemicals, stains and fine cracks | |
5°C to 40°C | |
51.8 cm | |
34.1 cm | |
41.4 cm |
Horizontal | |
Bottom-hinged, pull down oven style | |
Carrying handles, 2 latches and loops for a padlock or tamper-evident seals, door-mounted hygrometer | |
2 | |
41°F to 104°F | |
20.4 in. | |
13.4 in. | |
16.3 in. |