Learn More
SP Bel-Art™ Secador™ Standard Desiccator Cabinets
Constructed of Durastar™ co-polyester. Bel Art™ SP Scienceware™ Secador™ Standard Desiccator Cabinets: Vertical Models block 99% of UV light and are resistant to staining, crazing and chemical attack.
Brand: SP Bel-Art™ F42072-1008
Code : A0
Informations supplémentaires : Weight : 0.80000kg
Description
Constructed of Durastar™ co-polyester. Bel Art™ SP Scienceware™ Secador™ Standard Desiccator Cabinets: Vertical Models block 99% of UV light and are resistant to staining, crazing and chemical attack.
- Amber color models reduce visible light penetration by over 50% making it the perfect choice for storing light sensitive materials like reagents, analytical standards and investigational compounds
- Large doors maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals
- Perforated shelves facilitate circulation of dry air; space for desiccant under bottom shelf
- Easy-to-view digital hygrometer is located in the door front; side swinging door opens to the left
- Compact 34.1W x 41.4cmD footprint for space saving stacking up to 3 units high
- Replacement shelves available; 4.0 units can be converted from vertical to horizontal configuration or vice versa by turning the unit and ordering the appropriate number of shelves
- Can be used with desiccant cartridges, F42049-0100 or F42046-0000
Spécification
Not Autoclavable | |
Vertical | |
Left-hinged and gasketed | |
Carrying handles, 2 latches and loops for a padlock or tamper-evident seals, door-mounted hygrometer | |
2 | |
Removable Perforated | |
16.3 x 13.4 in. (41.4 x 34.1 cm) | |
41.4 cm | |
13.4 in. | |
34.1 cm | |
16.3 in. | |
16.3 in. |
Amber | |
Copolyester | |
Easy and reliable storage of moisture and light sensitive products, blocks 99% UV light, used with disposable or reusable desiccant | |
2.0 | |
Chemicals, stains and fine cracks | |
1.17 cu. ft. | |
16.3 in. | |
13.4 in. | |
34.1 cm | |
12.4 in. | |
31.4 cm | |
41.4 cm |