missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Spill Containment Tray with Grid

Large, deep, polypropylene tray protects users by capturing accidental hazardous spills

Brand:  Bel-Art™ F24676-0003

Product Code. 15152986

  • £117.00 / Each
Estimated Shipment: 27-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • The epoxy coated wire support grid holds numerous vessels or racks of test tubes
  • Outstanding chemical resistance
  • Steam autoclavable at 121°C (250°F)
Specifications

Specifications

236.69 oz.
Polypropylene
Bench top use
Grid
7 L
Spill Containment Tray
Multiple flasks, beakers, or a rack of test tubes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bel-Art™ Spill Containment Tray with Grid > 7L volume; 36.5 x 30.8 x 12.1cm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.