Learn More
Bel-Art™ Sterileware™ Scoop An' Bag™ Sterile Sampler
Sample collection system includes a sterile 60 mL (2 oz.) sampling scoop and sterile 11.4 x 22.9 cm (4.5 x 9 in. ), 540 mL (18.3 oz.) volume polyethylene twirl-type bag with large label area to hold the sample.
Brand:  Bel-Art™ H36910-0000
Product Code. 11741566
  • £123.00 / Pack of 50
Estimated Shipment: 02-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • One-piece scoop made from white, high-impact, FDA grade polystyrene plastic
  • Long handle keeps the hand away from the sample
  • Flat bottom allows scoop to remain upright for use as a weighing boat or to be placed on a flat surface
  • 50 individually sealed and sterilized wrapped sets per box ready for one time use and disposal sterile
  • Lot stamped for accurate lot tracing; shipped with a Certificate of Processing for Sterilization
Order Info
Sampling system comprised of a 2 oz. (60mL) white polystyrene scoop and a 9L x 4 0.50 in.W (228.6 x 114.3mm) polyethylene bag with a twirl-type closure packaged together.
Specifications
Specifications
Scoop Sampling System
Not Autoclavable
Sterile
Ideal for food, pharmaceutical and environmental sampling
High-impact Polystyrene (FDA Grade)
60mL white polystyrene flat bottom one-piece sampling scoop with 12.74cm long handle and 7.8 L x 4.1 W x 2.9cm H bowl, 540mL 11.4 x 22.9cm polyethylene twirl-type bag with large label area to hold the sample
FDA
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Bel-Art™ Sterileware™ Scoop An' Bag™ Sterile Sampler > Sterileware Scoop An'Bag, 2 oz.