Learn More
Köttermann Bench-type Hood
Designed for universal use, this Bench-type Hood is ideal for dealing with organic solvents and is also suitable for working with cold or diluted acids.
Brand:  Köttermann 451-00032
Additional Details : Weight : 73.35900kg
 View more versions of this product
Product Code. 17362189
  • £1559.79 / Each
Estimated Shipment: 05-11-2024
to see stock.
Description
Description
  • All-purpose hood for general use
  • Optimized protection from volatile hazardous materials
  • Long-lasting steel construction
  • Ergonomic features to improve workflow
Specifications
Specifications
No Ventilation Fan
GB Plug
85 kg
800 x 1350 x 640 mm
230 V