missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Köttermann Bench-type Hood
Designed for universal use, this Bench-type Hood is ideal for dealing with organic solvents and is also suitable for working with cold or diluted acids.
£1559.79 - £1773.54
Specifications
Dimensions (L x W x H) | 800 x 1350 x 640 mm |
---|---|
Voltage | 230 V |
Weight (Metric) | 85 kg |
Product Code | Brand | Configuration Options | Plug Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Configuration Options | Plug Type | Price | Quantity & Availability | ||||
17342189
|
Köttermann
451-00028 |
No Ventilation Fan | Plug Type D/F (Schuko/Old British Plug) |
£1571.34
Each |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17362189
|
Köttermann
451-00032 |
No Ventilation Fan | GB Plug |
£1559.79
Each |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17352189
|
Köttermann
451-00031 |
Ventilation Fan | GB Plug |
£1773.54
Each |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- All-purpose hood for general use
- Optimized protection from volatile hazardous materials
- Long-lasting steel construction
- Ergonomic features to improve workflow
Specifications
800 x 1350 x 640 mm | |
230 V | |
85 kg |