missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Benchtop Coolers
GREENER_CHOICE

Protect enzymes, cells, reagents and solutions by maintaining freezer temperatures while on the laboratory bench using benchtop coolers.

Brand:  Thermo Scientific™ 5116-1300

268.68 GBP valid until 2025-03-21
Use promo code "24111" to get your promotional price.


376.15 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10525021

  • £403.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Protect enzymes, cells, reagents and solutions by maintaining freezer temperatures while on the laboratory bench using Thermo Scientific™ Benchtop Coolers.

  • Maintain freezer temperature during frost-free cycles, power failures and accidental door openings
  • Holds 12, 20 or 32 tubes
  • Inserts are included for 1.5 mL tubes to have the compatibility to hold 0.2-0.5 mL microcentrifuge tubes
  • Printed grid on one-way keyed lid for proper orientation and sample identification
  • Molded of impact-resistant polycarbonate with locking handle for long-term use
  • Stackable to save space
TRUSTED_SUSTAINABILITY
Specifications

Specifications

190 mm
197 mm
140 mm
7.75 x 5.5 x 7.5 in. (197 x 140 x 190 mm)
3 x 4
Non-filled/clear lid
Benchtop Cooler
0.2 to 0.5 mL
0°C
7.5 in.
7.75 in.
5.5 in.
3x4 compartments, for 12-13mm dia.vials
<1°C, 5.0
Polycarbonate
Clear
12 Tubes (12 to 13 mm dia.)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1314388 Certificate TypeCertificate of Compliance Date10/12/2024 Catalog Number5116-1300
Lot Number1404520 Certificate TypeCertificate of Compliance Date22/04/2024 Catalog Number5116-1300
Lot Number1394460 Certificate TypeCertificate of Compliance Date28/01/2024 Catalog Number5116-1300
Lot Number1374641 Certificate TypeCertificate of Compliance Date30/11/2022 Catalog Number5116-1300
Lot Number1351194 Certificate TypeCertificate of Compliance Date10/03/2022 Catalog Number5116-1300
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Benchtop Coolers > Nalgene Labtop Cooler, 3x4 compartments, clear lid, accommodates 12 to 13mm dia. cryogenic vials

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.