missing translation for 'onlineSavingsMsg'
Learn More
Benzalkonium chloride, alkyl distribution from C8H17 to C16H33, Thermo Scientific Chemicals Product Code.: 10503771

Benzalkonium chloride, alkyl distribution from C8H17 to C16H33, Thermo Scientific Chemicals

Product Code. 10503771
100 g, Glass bottle
Click to view available options
Quantity:
100 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic Bottle
This item is not returnable. View return policy

Product Code. 10503771

Brand: Thermo Scientific Acros 215411000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8001-54-5
Molecular Formula C9H13ClN
Molecular Weight (g/mol) 170.66
MDL Number MFCD00145757
Synonym Alkyl-benzyl-dimethylammonium chloride

Specifications

Melting Point 40.0°C
Physical Form Solid or Thick Gel
Quantity 100 g
Linear Formula C6H5CH2N(CH3)2RClR=C8H17toC16H33
Merck Index 15, 1061
Solubility Information Solubility in water: 4000g/L (20°C). Other solubilities: soluble in alcohol and acetone,slightly soluble in benzene,almost insoluble in ether
Percent Purity 95.0 to 104.0% (anhydrous basis)
Infrared Spectrum Authentic
Packaging Glass bottle
Specific Gravity 0.98
Density 0.9800g/mL
Chemical Name or Material Benzalkonium chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467984 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number215410010,215411000,215410051
Lot NumberA0463365 Certificate TypeCertificate of Analysis Date14/06/2024 Catalog Number215410010,215411000,215410050,215410051
Lot NumberA0461338 Certificate TypeCertificate of Analysis Date25/04/2024 Catalog Number215410010,215411000,215410050,215410051
Lot NumberA0459817 Certificate TypeCertificate of Analysis Date06/02/2024 Catalog Number215410010,215411000,215410050,215410051
Lot NumberA0458735 Certificate TypeCertificate of Analysis Date18/12/2023 Catalog Number215410010,215411000,215410050,215410051
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Benzalkonium chloride, alkyl distribution from C8H17 to C16H36
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P262-Do not get in eyes, on skin, or on clothing.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P314-Get medical advice/attention if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Benzalkonium chloride, alkyl distribution from C8H17 to C16H33, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.