missing translation for 'onlineSavingsMsg'
Learn More

Benzenesulfonic acid, 90%, technical, Thermo Scientific Chemicals

£13.09 - £181.91

Chemical Identifiers

CAS 98-11-3, 7664-93-9
Molecular Formula C6H6O3S
Molecular Weight (g/mol) 158.17
MDL Number MFCD00011689
InChI Key SRSXLGNVWSONIS-UHFFFAOYSA-N
Synonym benzenesulphonic acid, phenylsulfonic acid, besylic acid, benzene sulphonic acid, benzenemonosulfonic acid, benzene sulfonic acid, kyselina benzensulfonova, benzensulfonic acid, ccris 4595, kyselina benzensulfonova czech
PubChem CID 7371
ChEBI CHEBI:64455
IUPAC Name benzenesulfonic acid
SMILES C1=CC=C(C=C1)S(=O)(=O)O
View More Specs

Products 6
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10406810
View Documents
Thermo Scientific Acros
159020050
5 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10076343
View Documents
Thermo Scientific Acros
159020250
25 g Glass bottle
£17.49
25g
Please to purchase this item. Need a web account? Register with us today!
10781355
View Documents
Thermo Scientific Acros
159021000
100 g Glass bottle
£18.79
100g
Please to purchase this item. Need a web account? Register with us today!
10187993
View Documents
Thermo Scientific Acros
159025000
500 g Glass bottle
£28.74
500g
Please to purchase this item. Need a web account? Register with us today!
10724651
View Documents
Thermo Scientific Acros
159020010
1 kg Glass bottle
£43.42
1kg
Please to purchase this item. Need a web account? Register with us today!
13486338
View Documents
Thermo Scientific Acros
159020051
5 kg Plastic drum
£181.91
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

98-11-3, 7664-93-9
158.17
SRSXLGNVWSONIS-UHFFFAOYSA-N
7371
benzenesulfonic acid
C6H6O3S
MFCD00011689
benzenesulphonic acid, phenylsulfonic acid, besylic acid, benzene sulphonic acid, benzenemonosulfonic acid, benzene sulfonic acid, kyselina benzensulfonova, benzensulfonic acid, ccris 4595, kyselina benzensulfonova czech
CHEBI:64455
C1=CC=C(C=C1)S(=O)(=O)O

Specifications

98-11-3, 7664-93-9
100.0
Yellow to Brown
Authentic
C6H6O3S
MFCD00011689
15,1071
Solubility in water: soluble. Other solubilities: soluble in alcohol,slightly soluble in benzene,insoluble in ether and carbon disulfide
C1=CC=C(C=C1)S(=O)(=O)O
158.17
CHEBI:64455
90%
Damp Crystalline Solid or Fused Mass
88.0
45.0°C to 50.0°C
>100°C
90%
C6H5SO3H
11,26
benzenesulphonic acid, phenylsulfonic acid, besylic acid, benzene sulphonic acid, benzenemonosulfonic acid, benzene sulfonic acid, kyselina benzensulfonova, benzensulfonic acid, ccris 4595, kyselina benzensulfonova czech
SRSXLGNVWSONIS-UHFFFAOYSA-N
benzenesulfonic acid
7371
158.17
Technical
Benzenesulfonic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 202-638-7

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.