missing translation for 'onlineSavingsMsg'
Learn More
Benzoic Acid, Certified AR for Analysis, Fisher Chemical™ Product Code.: 10476860

Benzoic Acid, Certified AR for Analysis, Fisher Chemical™

Product Code. 10476860
100 g
Click to view available options
Quantity:
100 g
500 g
1 kg
This item is not returnable. View return policy

Product Code. 10476860

Brand: Fisher Chemical B/1950/48

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Benzoic Acid, Certified AR for Analysis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65-85-0
Molecular Formula C7H6O2
MDL Number 2398

Specifications

Melting Point 121°C
Color White
pH 2.5 to 3.5
Boiling Point 249°C
Packaging Plastic powder jar
Quantity 100 g
Formula Weight 122.12g/mol
Physical Form Solid
compliance-icons compliance-icons
Product Identifier
  • Benzoic acid
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H372-Causes damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Benzoic Acid, Certified AR for Analysis, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.