Learn More
Benzoin, 98%, Thermo Scientific Chemicals
£18.09 - £141.71
Chemical Identifiers
CAS | 119-53-9 |
---|---|
Molecular Formula | C14H12O2 |
Molecular Weight (g/mol) | 212.25 |
MDL Number | MFCD00004496 |
InChI Key | ISAOCJYIOMOJEB-UHFFFAOYNA-N |
Synonym | benzoin, 2-hydroxy-2-phenylacetophenone, benzoylphenylcarbinol, benzoin tincture, +--benzoin, ethanone, 2-hydroxy-1,2-diphenyl, bitter almond oil camphor, phenylbenzoyl carbinol, 2-hydroxy-1,2-diphenylethan-1-one, alpha-hydroxybenzyl phenyl ketone |
PubChem CID | 8400 |
ChEBI | CHEBI:17682 |
IUPAC Name | 2-hydroxy-1,2-diphenylethanone |
SMILES | OC(C(=O)C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10457860
|
Thermo Scientific Acros
105511000 |
100 g | Plastic bottle |
£18.09
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10010550
|
Thermo Scientific Acros
105515000 |
500 g | Plastic bottle |
£33.87
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10530604
|
Thermo Scientific Acros
105510025 |
2.5 kg | Plastic bottle |
£141.71
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
119-53-9 | |
212.25 | |
ISAOCJYIOMOJEB-UHFFFAOYNA-N | |
8400 | |
2-hydroxy-1,2-diphenylethanone |
C14H12O2 | |
MFCD00004496 | |
benzoin, 2-hydroxy-2-phenylacetophenone, benzoylphenylcarbinol, benzoin tincture, +--benzoin, ethanone, 2-hydroxy-1,2-diphenyl, bitter almond oil camphor, phenylbenzoyl carbinol, 2-hydroxy-1,2-diphenylethan-1-one, alpha-hydroxybenzyl phenyl ketone | |
CHEBI:17682 | |
OC(C(=O)C1=CC=CC=C1)C1=CC=CC=C1 |
Specifications
119-53-9 | |
100.0 | |
White to Yellow | |
1% max. | |
97.5% min. (GC) | |
C6H5CH(OH)COC6H5 | |
15, 1095 | |
Solubility in water: sparingly soluble | |
OC(C(=O)C1=CC=CC=C1)C1=CC=CC=C1 | |
212.25 | |
CHEBI:17682 | |
98% | |
Benzoin, 98% |
97.5 | |
133.0°C to 137.0°C | |
194.0°C (12.0 mmHg) | |
Authentic | |
C14H12O2 | |
MFCD00004496 | |
benzoin, 2-hydroxy-2-phenylacetophenone, benzoylphenylcarbinol, benzoin tincture, +--benzoin, ethanone, 2-hydroxy-1,2-diphenyl, bitter almond oil camphor, phenylbenzoyl carbinol, 2-hydroxy-1,2-diphenylethan-1-one, alpha-hydroxybenzyl phenyl ketone | |
ISAOCJYIOMOJEB-UHFFFAOYNA-N | |
2-hydroxy-1,2-diphenylethanone | |
8400 | |
212.25 | |
Powder |
Safety and Handling
missing translation for 'einecsNumber' : 204-331-3
missing translation for 'rtecsNumber' : DI1590000
missing translation for 'tsca' : TSCA
- Benzoin
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.