missing translation for 'onlineSavingsMsg'
Learn More
Benzoin, 99%, Thermo Scientific Chemicals Product Code.: 11479492

Benzoin, 99%, Thermo Scientific Chemicals

Product Code. 11479492
1000 g
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy

Product Code. 11479492

Brand: Thermo Scientific Alfa Aesar A10188.0B

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Benzoin is used as a flavor, antiseptic and photopolymerization catalyst. It is also used as a raw material in organic syntheses. It serves as a precursor to benzil, which is a photoinitiator. Further, it is used as degassing agent for powder coatings. In addition to this, it is used as an additive and in powder coating to remove the pinhole phenomenon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzoin is used as a flavor, antiseptic and photopolymerization catalyst. It is also used as a raw material in organic syntheses. It serves as a precursor to benzil, which is a photoinitiator. Further, it is used as degassing agent for powder coatings. In addition to this, it is used as an additive and in powder coating to remove the pinhole phenomenon.

Solubility
Soluble in chlorine. Slightly soluble in water, ethanol and ether.

Notes
Moisture sensitive. Incompatible with acids, acid anhydrides, acid chlorides and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-53-9
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.25
MDL Number MFCD00004496
InChI Key ISAOCJYIOMOJEB-UHFFFAOYNA-N
Synonym benzoin, 2-hydroxy-2-phenylacetophenone, benzoylphenylcarbinol, benzoin tincture, +--benzoin, ethanone, 2-hydroxy-1,2-diphenyl, bitter almond oil camphor, phenylbenzoyl carbinol, 2-hydroxy-1,2-diphenylethan-1-one, alpha-hydroxybenzyl phenyl ketoneShow MoreShow Less
PubChem CID 8400
ChEBI CHEBI:17682
IUPAC Name 2-hydroxy-1,2-diphenylethanone
SMILES OC(C(=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 131°C to 136°C
Density 1.31
Boiling Point 334°C to 344°C
Flash Point 181°C (357°F)
Odor Pleasant
Quantity 1000 g
Beilstein 391839
Merck Index 14,1093
Solubility Information Soluble in chlorine. Slightly soluble in water,ethanol and ether.
Formula Weight 212.25
Percent Purity 99%
Chemical Name or Material Benzoin
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10243379 Certificate TypeCertificate of Analysis Date26/09/2022 Catalog NumberA10188.0I,A10188.30,A10188.0B
Lot Number10238587 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA10188.0I,A10188.30,A10188.0B
Lot NumberH7859A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10188.0I,A10188.30,A10188.0B
Lot Number10195778 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10188.0I,A10188.30,A10188.0B
Lot Number10149529 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10188.0I,A10188.30,A10188.0B

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Benzoin, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.