Learn More
Benzoylacetone, 98+%, Thermo Scientific Chemicals
£35.70 - £438.00
Chemical Identifiers
CAS | 93-91-4 |
---|---|
Molecular Formula | C10H10O2 |
Molecular Weight (g/mol) | 162.188 |
MDL Number | MFCD00008786 |
InChI Key | CVBUKMMMRLOKQR-UHFFFAOYSA-N |
Synonym | benzoylacetone, 1-phenyl-1,3-butanedione, 1-benzoylacetone, 1,3-butanedione, 1-phenyl, 1-benzoyl-2-propanone, acetoacetophenone, 2-propanone, benzoyl, acetylbenzoylmethane, 2-acetylacetophenone, benzoyl-aceton |
PubChem CID | 7166 |
IUPAC Name | 1-phenylbutane-1,3-dione |
SMILES | CC(=O)CC(=O)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11411908
|
Thermo Scientific Alfa Aesar
A14537.18 |
50 g |
£35.70
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11421908
|
Thermo Scientific Alfa Aesar
A14537.30 |
250 g |
£138.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11401908
|
Thermo Scientific Alfa Aesar
A14537.0B |
1000 g |
£438.00
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Benzoylacetone has been used as a 1,3-dicarbonyl compound model for studying keto-enol equilibria in aqueous acid and micellar solutions. Additionally, it has been studied using gas-phase electron diffraction and quantum chemistry. It is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBenzoylacetone has been used as a 1,3-dicarbonyl compound model for studying keto-enol equilibria in aqueous acid and micellar solutions. Additionally, it has been studied using gas-phase electron diffraction and quantum chemistry. It is used as pharmaceutical intermediates.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
93-91-4 | |
162.188 | |
CVBUKMMMRLOKQR-UHFFFAOYSA-N | |
7166 | |
CC(=O)CC(=O)C1=CC=CC=C1 |
C10H10O2 | |
MFCD00008786 | |
benzoylacetone, 1-phenyl-1,3-butanedione, 1-benzoylacetone, 1,3-butanedione, 1-phenyl, 1-benzoyl-2-propanone, acetoacetophenone, 2-propanone, benzoyl, acetylbenzoylmethane, 2-acetylacetophenone, benzoyl-aceton | |
1-phenylbutane-1,3-dione |
Specifications
93-91-4 | |
1.09 | |
C10H10O2 | |
742413 | |
Insoluble in water. | |
CC(=O)CC(=O)C1=CC=CC=C1 | |
162.188 | |
162.19 | |
Benzoylacetone |
55°C to 60°C | |
260°C to 262°C | |
MFCD00008786 | |
benzoylacetone, 1-phenyl-1,3-butanedione, 1-benzoylacetone, 1,3-butanedione, 1-phenyl, 1-benzoyl-2-propanone, acetoacetophenone, 2-propanone, benzoyl, acetylbenzoylmethane, 2-acetylacetophenone, benzoyl-aceton | |
CVBUKMMMRLOKQR-UHFFFAOYSA-N | |
1-phenylbutane-1,3-dione | |
7166 | |
≥98% |
Safety and Handling
missing translation for 'einecsNumber' : 202-286-4
missing translation for 'rtecsNumber' : EK3540200
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only