missing translation for 'onlineSavingsMsg'
Learn More
Benzyl Alcohol, puriss., 99 to 100.5% (GC), Honeywell Riedel-de Haën™ Product Code.: 15632500

Benzyl Alcohol, puriss., 99 to 100.5% (GC), Honeywell Riedel-de Haën™
BIOPHARMA

Product Code. 15632500
4 x 2.5 L, Glass Bottle
Click to view available options
Quantity:
4 x 2.5 L
6 x 1 L
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 15632500

Brand: Honeywell-Riedel-de Haen 241224X2.5L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss., meets analytical specification of BP, Ph. Eur., NF, 99-100.5% (GC)

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-51-6
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00004599,MFCD03792087
InChI Key WVDDGKGOMKODPV-UHFFFAOYSA-N
Synonym benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcoholShow MoreShow Less
PubChem CID 244
ChEBI CHEBI:17987
IUPAC Name phenylmethanol
SMILES OCC1=CC=CC=C1

Specifications

Boiling Range 203° to 205°C (lit.)
Melting Point -16°C to -13°C
Density 1.045g/mL at 25°C
Flash Point 96°C (204.8°F)
Ignition Residue ≤0.005% (as SO4)
Ignition Point 817°F
Packaging Glass Bottle
Linear Formula C6H5CH2OH
Quantity 4 x 2.5 L
UN Number UN3334
Beilstein 878307
Vapor Density 3.7 (vs air)
Formula Weight 108.14g/mol
Vapor Pressure 13.3mmHg (100°C), 3.75mmHg (77°C)
Percent Purity 99 to 100.5% (GC)
Grade puriss.
Physical Form Liquid
Chemical Name or Material Benzyl Alcohol, Meets analytical specification of Ph.Eur.BP, NF
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM3270 Certificate TypeCertificate of Analysis Date05/04/2023 Catalog Number24122-1L,24122-2.5L,24122-30KG,24122-4X2.5L,24122-6X1L
compliance-icons
Product Identifier
  • Benzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302 + H332-Harmful if swallowed or if inhaled.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Benzyl Alcohol, puriss., 99 to 100.5% (GC), Honeywell Riedel-de Haën™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.