missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl ethyl malonate, tech. 85%, Thermo Scientific Chemicals
£107.00 - £1382.00
Chemical Identifiers
CAS | 42998-51-6 |
---|---|
Molecular Formula | C12H14O4 |
Molecular Weight (g/mol) | 222.24 |
MDL Number | MFCD00009194 |
InChI Key | CGNOCUSLPSCMLL-UHFFFAOYSA-N |
Synonym | benzyl ethyl malonate, benzylethyl malonate, malonic acid benzyl ethyl ester, ethyl benzylmalonate, acmc-20alpf, 1-benzyl 3-ethyl malonate #, ethyl phenylmethyl propanedioate, 1-benzyl 3-ethyl propanedioate, malonic acid 1-benzyl 3-ethyl ester, ethyl phenylmethyl propane-1,3-dioate |
PubChem CID | 562228 |
SMILES | CCOC(=O)CC(=O)OCC1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11446186
|
Thermo Scientific Alfa Aesar
B23011.06 |
5 g |
£107.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11456186
|
Thermo Scientific Alfa Aesar
B23011.14 |
25 g |
£421.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11466186
|
Thermo Scientific Alfa Aesar
B23011.22 |
100 g |
£1382.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Benzyl ethyl malonate is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBenzyl ethyl malonate is used as a pharmaceutical intermediate.
Solubility
Soluble in water 525.7 mg/L at 25°C. Soluble in alcohol.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Chemical Identifiers
42998-51-6 | |
222.24 | |
CGNOCUSLPSCMLL-UHFFFAOYSA-N | |
562228 |
C12H14O4 | |
MFCD00009194 | |
benzyl ethyl malonate, benzylethyl malonate, malonic acid benzyl ethyl ester, ethyl benzylmalonate, acmc-20alpf, 1-benzyl 3-ethyl malonate #, ethyl phenylmethyl propanedioate, 1-benzyl 3-ethyl propanedioate, malonic acid 1-benzyl 3-ethyl ester, ethyl phenylmethyl propane-1,3-dioate | |
CCOC(=O)CC(=O)OCC1=CC=CC=C1 |
Specifications
42998-51-6 | |
C12H14O4 | |
MFCD00009194 | |
Soluble in water 525.7mg/L @ 25°C. Soluble in alcohol. | |
CCOC(=O)CC(=O)OCC1=CC=CC=C1 | |
222.24 | |
222.24 | |
Technical |
145°C (4 mmHg) | |
1.497 | |
benzyl ethyl malonate, benzylethyl malonate, malonic acid benzyl ethyl ester, ethyl benzylmalonate, acmc-20alpf, 1-benzyl 3-ethyl malonate #, ethyl phenylmethyl propanedioate, 1-benzyl 3-ethyl propanedioate, malonic acid 1-benzyl 3-ethyl ester, ethyl phenylmethyl propane-1,3-dioate | |
CGNOCUSLPSCMLL-UHFFFAOYSA-N | |
3-O-benzyl 1-O-ethyl propanedioate | |
562228 | |
85% | |
Benzyl ethyl malonate |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only