Learn More
Benzylamine, 98+%, Thermo Scientific Chemicals
£32.26 - £345.72
Chemical Identifiers
CAS | 100-46-9 |
---|---|
Molecular Formula | C7H9N |
Molecular Weight (g/mol) | 107.156 |
MDL Number | MFCD00008106 |
InChI Key | WGQKYBSKWIADBV-UHFFFAOYSA-N |
Synonym | benzylamine, benzenemethanamine, monobenzylamine, alpha-aminotoluene, phenylmethyl amine, aminomethyl benzene, 1-phenylmethanamine, moringine, n-benzylamine, phenylmethylamine |
PubChem CID | 7504 |
ChEBI | CHEBI:40538 |
IUPAC Name | phenylmethanamine |
SMILES | C1=CC=C(C=C1)CN |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11474443
|
Thermo Scientific Alfa Aesar
A10997.30 |
250 g |
£32.26
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11364579
|
Thermo Scientific Alfa Aesar
|
500 g |
£54.27
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11464443
|
Thermo Scientific Alfa Aesar
A10997.0B |
1000 g |
£88.04
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11484443
|
Thermo Scientific Alfa Aesar
A10997.0I |
5000 g |
£345.72
5000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Benzylamine is a valuable intermediate for various applications and a building block for chemical synthesis used in pharmaceuticals and crop protection agents. It finds application in the coating industry. It is involved as a carrier electrolyte for separation of alkali, alkaline earth and ammonium cations in water samples by capillary electrophoresis with indirect UV detection. It is also used in synthesis of cross-linked porous copolymer supports based on N-(p-vinylbenzoyl)-2-methylalanine, styrene and divinylbenzene. In the textile industry, it is used in colored dyes. It is often used for medicinal purposes in topical creams and antifungal solutions as well as in vitamins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBenzylamine is a valuable intermediate for various applications and a building block for chemical synthesis used in pharmaceuticals and crop protection agents. It finds application in the coating industry. It is involved as a carrier electrolyte for separation of alkali, alkaline earth and ammonium cations in water samples by capillary electrophoresis with indirect UV detection. It is also used in synthesis of cross-linked porous copolymer supports based on N-(p-vinylbenzoyl)-2-methylalanine, styrene and divinylbenzene. In the textile industry, it is used in colored dyes. It is often used for medicinal purposes in topical creams and antifungal solutions as well as in vitamins.
Solubility
Miscible with water, ethanol, diethyl ether, acetone, benzene and chloroform.
Notes
Air sensitive. Incompatible with strong oxidizing agents and strong acids.
Chemical Identifiers
100-46-9 | |
107.156 | |
WGQKYBSKWIADBV-UHFFFAOYSA-N | |
7504 | |
phenylmethanamine |
C7H9N | |
MFCD00008106 | |
benzylamine, benzenemethanamine, monobenzylamine, alpha-aminotoluene, phenylmethyl amine, aminomethyl benzene, 1-phenylmethanamine, moringine, n-benzylamine, phenylmethylamine | |
CHEBI:40538 | |
C1=CC=C(C=C1)CN |
Specifications
100-46-9 | |
0.982 | |
72°C (162°F) | |
1.5435 | |
UN2735 | |
Air Sensitive | |
benzylamine, benzenemethanamine, monobenzylamine, alpha-aminotoluene, phenylmethyl amine, aminomethyl benzene, 1-phenylmethanamine, moringine, n-benzylamine, phenylmethylamine | |
WGQKYBSKWIADBV-UHFFFAOYSA-N | |
phenylmethanamine | |
7504 | |
107.16 | |
Benzylamine |
-43°C | |
184°C to 185°C | |
C7H9N | |
MFCD00008106 | |
741984 | |
14,1125 | |
Miscible with water,ethanol,diethyl ether,acetone,benzene and chloroform. | |
C1=CC=C(C=C1)CN | |
107.156 | |
CHEBI:40538 | |
≥98% |
Safety and Handling
GHS H Statement
H314-H302-H312-H227
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Combustible liquid.
P210-P233-P240-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P363-P374-P501c
H227-H302+H312-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
missing translation for 'einecsNumber' : 202-854-1
missing translation for 'rtecsNumber' : DP1488500
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
- Benzylamine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Benzenemethanamine
RUO – Research Use Only