missing translation for 'onlineSavingsMsg'
Learn More
Benzylamine, 98+%, Thermo Scientific Chemicals Product Code.: 11474443

Benzylamine, 98+%, Thermo Scientific Chemicals

Product Code. 11474443
250 g
Click to view available options
Quantity:
250 g
500 g
1000 g
5000 g
This item is not returnable. View return policy

Product Code. 11474443

Brand: Thermo Scientific Alfa Aesar A10997.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Benzylamine is a valuable intermediate for various applications and a building block for chemical synthesis used in pharmaceuticals and crop protection agents. It finds application in the coating industry. It is involved as a carrier electrolyte for separation of alkali, alkaline earth and ammonium cations in water samples by capillary electrophoresis with indirect UV detection. It is also used in synthesis of cross-linked porous copolymer supports based on N-(p-vinylbenzoyl)-2-methylalanine, styrene and divinylbenzene. In the textile industry, it is used in colored dyes. It is often used for medicinal purposes in topical creams and antifungal solutions as well as in vitamins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzylamine is a valuable intermediate for various applications and a building block for chemical synthesis used in pharmaceuticals and crop protection agents. It finds application in the coating industry. It is involved as a carrier electrolyte for separation of alkali, alkaline earth and ammonium cations in water samples by capillary electrophoresis with indirect UV detection. It is also used in synthesis of cross-linked porous copolymer supports based on N-(p-vinylbenzoyl)-2-methylalanine, styrene and divinylbenzene. In the textile industry, it is used in colored dyes. It is often used for medicinal purposes in topical creams and antifungal solutions as well as in vitamins.

Solubility
Miscible with water, ethanol, diethyl ether, acetone, benzene and chloroform.

Notes
Air sensitive. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-46-9
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.156
MDL Number MFCD00008106
InChI Key WGQKYBSKWIADBV-UHFFFAOYSA-N
Synonym benzylamine, benzenemethanamine, monobenzylamine, alpha-aminotoluene, phenylmethyl amine, aminomethyl benzene, 1-phenylmethanamine, moringine, n-benzylamine, phenylmethylamineShow MoreShow Less
PubChem CID 7504
ChEBI CHEBI:40538
IUPAC Name phenylmethanamine
SMILES C1=CC=C(C=C1)CN

Specifications

Melting Point -43°C
Density 0.982
Boiling Point 184°C to 185°C
Flash Point 72°C (162°F)
Refractive Index 1.5435
Quantity 250 g
UN Number UN2735
Beilstein 741984
Sensitivity Air Sensitive
Merck Index 14,1125
Solubility Information Miscible with water,ethanol,diethyl ether,acetone,benzene and chloroform.
Formula Weight 107.16
Percent Purity ≥98%
Chemical Name or Material Benzylamine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10249774 Certificate TypeCertificate of Analysis Date05/03/2024 Catalog NumberA10997.30,A10997.0I,A10997.36,A10997.0B
Lot Number10247444 Certificate TypeCertificate of Analysis Date27/07/2023 Catalog NumberA10997.30,A10997.0I,A10997.36,A10997.0B
Lot Number61000169 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10997.30,A10997.0I,A10997.36,A10997.0B
Lot Number61201092 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10997.30,A10997.0I,A10997.36,A10997.0B
Lot NumberK06N02 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10997.30,A10997.0I,A10997.36,A10997.0B
compliance-icons compliance-icons
Product Identifier
  • Benzylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Benzenemethanamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Benzylamine, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.