missing translation for 'onlineSavingsMsg'
Learn More
Benzylmagnesium chloride, 1.5M solution in THF, Thermo Scientific Chemicals Product Code.: 10095093

Benzylmagnesium chloride, 1.5M solution in THF, Thermo Scientific Chemicals

Product Code. 10095093
100 mL, AcroSeal™ Glass bottle
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

Product Code. 10095093

Brand: Thermo Scientific Acros 389551000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6921-34-2, 109-99-9
Molecular Formula C7H7ClMg
Molecular Weight (g/mol) 150.89
MDL Number MFCD00000469
InChI Key NRAFPLGJPPJUNB-UHFFFAOYSA-M
Synonym benzylmagnesium chloride, magnesium, chloro phenylmethyl, phch2mgcl, benzyl chloro magnesium, benzyl magnesium chloride, chloro phenylmethyl magnesium, benzylmagnesiumchloride, bnmgcl, benzyl-magnesium chloride, chloro phenylmethyl-magnesiumShow MoreShow Less
PubChem CID 2733352
IUPAC Name benzyl(chloro)magnesium
SMILES Cl[Mg]CC1=CC=CC=C1

Specifications

Color Brown to Black
CAS Min % 20.51, 75.83
CAS Max % 24.17, 79.49
Linear Formula (C6H5)CH2MgCl
Concentration 1.40 to 1.65M
Density 1.0300g/mL
Solubility Information Solubility in water: reacts.
Formula Weight 150.89
Percent Purity 19 to 21% (as BMC) (Total base)
Packaging AcroSeal™ Glass bottle
Specific Gravity 1.03
Quantity 100 mL
Flash Point −17°C
Physical Form Iridescent Powder or Crystals
Chemical Name or Material Benzylmagnesium chloride, 1.5M solution in THF
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464588 Certificate TypeCertificate of Analysis Date24/07/2024 Catalog Number389551000
Lot NumberA0440564 Certificate TypeCertificate of Analysis Date16/05/2024 Catalog Number389551000
Lot NumberA0440564 Certificate TypeCertificate of Analysis Date22/04/2022 Catalog Number
Lot NumberA0429530 Certificate TypeCertificate of Analysis Date12/05/2021 Catalog Number389558000,389551000
Lot NumberA0403214 Certificate TypeCertificate of Analysis Date07/01/2021 Catalog Number389558000,389551000
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Benzylmagnesium chloride, 1-2M solution in THF
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Tetrahydrofuran, Benzylmagnesium chloride
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138

    Expires2025-04-30

    Days

    30

    Hours

    6

    Minutes

    30

    Seconds

    16

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Benzylmagnesium chloride, 1.5M solution in THF, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.