missing translation for 'onlineSavingsMsg'
Learn More
Benzyltrimethylammonium bromide, 98+%, Thermo Scientific Chemicals Product Code.: 11452868

Benzyltrimethylammonium bromide, 98+%, Thermo Scientific Chemicals

Product Code. 11452868
100 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11452868

Brand: Thermo Scientific Alfa Aesar A16840.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5350-41-4
Molecular Formula C10H16BrN
Molecular Weight (g/mol) 230.149
MDL Number MFCD00011780
InChI Key UUZYBYIOAZTMGC-UHFFFAOYSA-M
Synonym benzyltrimethylammonium bromide, n,n,n-trimethyl-1-phenylmethanaminium bromide, trimethylbenzylammonium bromide, n-benzyl-n,n,n-trimethylammonium bromide, wv 562 german, benzenemethanaminium, n,n,n-trimethyl-, bromide, nsc 24, ammonium, benzyltrimethyl-, bromide, benzyltrimethylammonium bromide btmShow MoreShow Less
PubChem CID 21449
IUPAC Name benzyl(trimethyl)azanium;bromide
SMILES C[N+](C)(C)CC1=CC=CC=C1.[Br-]

Specifications

Melting Point 228°C to 232°C
Quantity 100 g
Beilstein 3917251
Sensitivity Hygroscopic
Formula Weight 230.15
Percent Purity ≥98%
Chemical Name or Material Benzyltrimethylammonium bromide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10117921 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16840.22,A16840.14
Lot Number10117921 Certificate TypeCertificate of Analysis Date07/08/2020 Catalog NumberA16840.22,A16840.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Benzyltrimethylammonium bromide, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.